siRNA Details
Gene Symbol: OR2G6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013355.1 | NP_001013373.1 | 1 | 637 | ||
GenBank mRNA | BC137245.1 | - | 1 | 669 | 9021242 | |
BC137266.1 | - | 1 | 669 | 9021263 |
