siRNA Details
Gene Symbol: OR13C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004481.1 | NP_001004481.1 | 1 | 828 | ||
GenBank mRNA | BC136824.1 | - | 1 | 871 | 9020815 | |
BC136826.1 | - | 1 | 871 | 9020817 |
Gene Symbol: OR13C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001956.1 | NP_001001956.1 | 1 | 828 |
