siRNA Details
Gene Symbol: OR1L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004453.2 | NP_001004453.2 | 1 | 756 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR1L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004453.2 | NP_001004453.2 | 1 | 756 |