siRNA Details
Gene Symbol: SMIM10L2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024493.2 | 2 | 767 | |||
GenBank mRNA | AK123302.1 | - | 2 | 767 |
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 964 | |||
GenBank mRNA | BC030620.1 | - | 2 | 963 | ||
BC051704.1 | - | 2 | 951 |
