siRNA Details
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 1331 | |||
GenBank mRNA | BC030620.1 | - | 2 | 1330 | ||
BC051704.1 | - | 2 | 1318 | |||
DR001138.1 | - | 1 | 350 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 1331 | |||
GenBank mRNA | BC030620.1 | - | 2 | 1330 | ||
BC051704.1 | - | 2 | 1318 | |||
DR001138.1 | - | 1 | 350 |