siRNA Details
Gene Symbol: GREB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014668.3 | NP_055483.2 | 16, 17 | 2833 | ||
XM_005246192.4 | XP_005246249.1 | 16, 17 | 2855 | |||
XM_011510418.2 | XP_011508720.1 | 15, 16 | 2694 | |||
XM_011510419.2 | XP_011508721.1 | 15, 16 | 2691 | |||
GenBank mRNA | AF245388.1 | - | 16, 17 | 2833 | ||
BC034455.1 | - | 7, 8 | 1864 |
