siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 86 | 15507 | ||
XM_006721206.2 | XP_006721269.1 | 86 | 15401 | |||
XM_011523146.2 | XP_011521448.1 | 87 | 15536 | |||
XM_011523147.1 | XP_011521449.1 | 87 | 15503 | |||
XM_011523148.1 | XP_011521450.1 | 87 | 15576 | |||
XM_011523151.1 | XP_011521453.1 | 86 | 15433 | |||
XM_011523152.1 | XP_011521454.1 | 49 | 9260 | |||
XM_011523155.2 | XP_011521457.1 | 42 | 8261 | |||
XM_017023346.1 | XP_016878835.1 | 86 | 15679 | |||
XM_017023347.1 | XP_016878836.1 | 74 | 13918 | |||
XM_017023348.1 | XP_016878837.1 | 74 | 13658 | |||
GenBank mRNA | AK026688.1 | - | 7 | 1603 | ||
AK074472.1 | - | 11 | 2326 | |||
AL137259.1 | - | 14 | 2903 | |||
BC043273.2 | - | 22 | 4351 |
