siRNA Details
Gene Symbol: C11orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207645.3 | NP_997528.2 | 2 | 670 | ||
XM_011542817.2 | XP_011541119.1 | 2 | 774 | |||
XM_011542818.2 | XP_011541120.1 | 2 | 652 | |||
GenBank mRNA | AK303693.1 | - | 2 | 670 | ||
BC035798.1 | - | 2 | 590 | 5735100 | ||
BC068577.1 | - | 2 | 643 | 4818568 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001286641, NM_178906, BC109328, AK033979.
Gene Ontology Categories:
