siRNA Details
Gene Symbol: C11orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100388.1 | NP_001093858.1 | 2, 3 | 275 | ||
NM_207430.2 | NP_997313.2 | 2, 3 | 275 | |||
GenBank mRNA | AK128145.1 | - | 2, 3 | 275 | ||
BC039505.2 | - | 2, 3 | 260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
