siRNA Details
Gene Symbol: CD3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000733.3 | NP_000724.1 | 7 | 629 | ||
GenBank mRNA | AK292612.1 | - | 7 | 478 | ||
AK311641.1 | - | 6 | 926 | |||
BC049847.1 | - | 6 | 441 | 5214563 | ||
BI910359.1 | - | 7 | 665 | |||
X03884.1 | - | 6 | 427 |
