siRNA Details
Gene Symbol: PLEKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161498.1 | NP_001154970.1 | 2 | 421 | ||
XM_017021290.1 | XP_016876779.1 | 2 | 783 | |||
GenBank mRNA | AK126770.1 | - | 2 | 483 | ||
BC146872.1 | - | 2 | 326 | 9056800 |
