siRNA Details
Gene Symbol: C15orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207444.2 | NP_997327.1 | 1, 2 | 151 | ||
GenBank mRNA | AK093014.1 | - | 1, 2 | 151 | ||
BC119016.1 | - | 1, 2 | 151 | 40108911 | ||
BC119810.1 | - | 1, 2 | 151 | 40108916 |
