siRNA Details
Gene Symbol: GRAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024637.1 | XP_016880126.1 | 5 | 1213 | ||
GenBank mRNA | BC026233.1 | - | 3 | 822 | ||
BC060883.1 | - | 3 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
