siRNA Details
Gene Symbol: ASXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030632.2 | NP_085135.1 | 11 | 2020 | ||
XM_005258356.1 | XP_005258413.1 | 12 | 1993 | |||
XM_011526205.2 | XP_011524507.1 | 12 | 8783 | |||
XM_011526206.2 | XP_011524508.1 | 12 | 5548 | |||
XM_011526209.1 | XP_011524511.1 | 13 | 2177 | |||
XM_011526210.1 | XP_011524512.1 | 13 | 2234 | |||
XM_011526212.1 | XP_011524514.1 | 12 | 2138 | |||
XM_011526213.1 | XP_011524515.1 | 12 | 2231 | |||
XM_017026012.1 | XP_016881501.1 | 12 | 5395 | |||
XM_017026013.1 | XP_016881502.1 | 13 | 5757 | |||
XM_017026014.1 | XP_016881503.1 | 13 | 2194 | |||
GenBank mRNA | AB051500.2 | - | 1 | 179 | ||
AK131454.1 | - | 13 | 2063 |
