siRNA Details
Gene Symbol: THSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018676.3 | NP_061146.1 | 5 | 2153 | ||
NM_199263.2 | NP_954872.1 | 4 | 1994 | |||
GenBank mRNA | AB044385.1 | - | 5 | 1759 | ||
AK096289.1 | - | 4 | 1994 | |||
AK298974.1 | - | 3 | 700 | |||
AK315087.1 | - | 5 | 1802 | |||
BC063842.1 | - | 4 | 1627 | 6181100 |
