siRNA Details
Gene Symbol: PRSS57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358594.1 | - | 3 | 390 | ||
BC137161.1 | - | 3 | 355 | 9021158 | ||
BC144505.1 | - | 3 | 352 | 9053032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
