siRNA Details
Gene Symbol: LOC400682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528477.2 | XP_011526779.2 | 4 | 1001 | ||
GenBank mRNA | BX647610.1 | - | 2 | 804 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC400682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528477.2 | XP_011526779.2 | 4 | 1001 | ||
GenBank mRNA | BX647610.1 | - | 2 | 804 |