siRNA Details
Gene Symbol: C1orf226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085375.1 | NP_001078844.1 | 1, 2 | 483 | ||
NM_001135240.1 | NP_001128712.1 | 2, 3 | 448 | |||
GenBank mRNA | AK125122.1 | - | 1, 2 | 483 | ||
AK293907.1 | - | 2, 3 | 448 | |||
BC127743.1 | - | 1, 2 | 353 | 40127716 | ||
BC127744.1 | - | 1, 2 | 350 | 40127717 |
