siRNA Details
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017547.3 | NP_060017.1 | 7 | 953 | ||
NR_037647.1 | 6 | 732 | ||||
NR_037648.1 | 7 | 972 | ||||
XM_006718879.3 | XP_006718942.1 | 6 | 666 | |||
XM_006718881.3 | XP_006718944.1 | 5 | 634 | |||
XM_011542895.2 | XP_011541197.1 | 7 | 1050 | |||
XM_011542896.2 | XP_011541198.1 | 7 | 859 | |||
XM_017018000.1 | XP_016873489.1 | 7 | 928 | |||
XM_017018001.1 | XP_016873490.1 | 7 | 1072 | |||
XM_017018002.1 | XP_016873491.1 | 6 | 581 | |||
XM_017018003.1 | XP_016873492.1 | 6 | 833 | |||
XM_017018004.1 | XP_016873493.1 | 6 | 955 | |||
XM_017018005.1 | XP_016873494.1 | 5 | 2188 | |||
XM_017018006.1 | XP_016873495.1 | 6 | 833 | |||
GenBank mRNA | AF103801.1 | - | 7 | 851 | ||
AF447877.1 | - | 5 | 1105 | |||
AK023987.1 | - | 7 | 822 | |||
AK126067.1 | - | 1 | 1931 | |||
AK295267.1 | - | 7 | 908 | |||
AK295428.1 | - | 7 | 838 | |||
AK298010.1 | - | 5 | 506 | |||
AK298807.1 | - | 7 | 838 | |||
AK302039.1 | - | 6 | 601 | |||
AL136923.1 | - | 7 | 833 | |||
BC002910.2 | - | 5 | 430 | |||
BC013902.2 | - | 7 | 809 | 3956972 |
