siRNA Details
Gene Symbol: RSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286506.1 | NP_001273435.1 | 4 | 542 | ||
NM_080860.3 | NP_543136.1 | 5 | 656 | |||
XM_005261208.2 | XP_005261265.1 | 3 | 296 | |||
XM_011529786.1 | XP_011528088.1 | 5 | 548 | |||
GenBank mRNA | AB006536.1 | - | 5 | 656 | ||
AB041016.1 | - | 5 | 593 | |||
AK057315.1 | - | 4 | 1115 | |||
AK314746.1 | - | 5 | 493 | |||
BC101519.1 | - | 5 | 535 | 8069025 | ||
BC113367.1 | - | 5 | 535 | 8322419 |
