siRNA Details
Gene Symbol: C10orf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305581.1 | NP_001292510.1 | 7 | 702 | ||
NM_032024.4 | NP_114413.1 | 6 | 754 | |||
NR_131178.1 | 8 | 977 | ||||
XM_011540257.2 | XP_011538559.1 | 6 | 611 | |||
XM_017016759.1 | XP_016872248.1 | 8 | 842 | |||
XM_017016760.1 | XP_016872249.1 | 8 | 743 | |||
XM_017016762.1 | XP_016872251.1 | 4 | 678 | |||
GenBank mRNA | AF267860.1 | - | 6 | 755 | ||
AK125328.1 | - | 3 | 985 | |||
BX105111.1 | - | 6 | 596 | |||
CR933639.1 | - | 8 | 977 |
