siRNA Details
Gene Symbol: LINC00898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033377.1 | 4 | 2619 | |||
GenBank mRNA | AK128136.1 | - | 4 | 2616 | ||
BC132762.1 | - | 4 | 952 | |||
BC132788.1 | - | 4 | 952 | |||
BC144501.1 | - | 3 | 871 | |||
BC144502.1 | - | 3 | 1000 |
