siRNA Details
Gene Symbol: FOSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006732.2 | NP_006723.2 | 3 | 1083 | ||
XM_005258691.1 | XP_005258748.1 | 3 | 1082 | |||
GenBank mRNA | AK225070.1 | - | 2 | 1327 | ||
AK292720.1 | - | 4 | 682 | |||
BC036724.1 | - | 4 | 1062 | 5212854 | ||
BC040197.1 | - | 2 | 1085 | 4816584 | ||
BX538117.1 | - | 3 | 1367 | |||
CR542101.1 | - | 4 | 491 | |||
EU178110.1 | - | 4 | 1084 | |||
EU178111.1 | - | 3 | 967 | |||
EU178114.1 | - | 3 | 967 | |||
L49169.1 | - | 4 | 1084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
