siRNA Details
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 9 | 1174 | ||
XM_011532845.2 | XP_011531147.1 | 10 | 1373 | |||
XM_011532849.2 | XP_011531151.1 | 10 | 1373 | |||
XM_011532850.1 | XP_011531152.1 | 10 | 1373 | |||
XM_011532851.2 | XP_011531153.1 | 10 | 1373 | |||
XM_011532852.2 | XP_011531154.1 | 10 | 1373 | |||
XM_017004103.1 | XP_016859592.1 | 10 | 1373 | |||
XM_017004104.1 | XP_016859593.1 | 9 | 1490 | |||
XM_017004105.1 | XP_016859594.1 | 9 | 1338 | |||
XM_017004106.1 | XP_016859595.1 | 9 | 1457 | |||
XM_017004107.1 | XP_016859596.1 | 10 | 1373 | |||
GenBank mRNA | AF237700.2 | - | 1 | 123 | ||
CR749330.1 | - | 9 | 1174 |
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 9 | 1220 | ||
XM_017004845.1 | XP_016860334.1 | 13 | 1671 | |||
XM_017004846.1 | XP_016860335.1 | 9 | 1342 | |||
XM_017004847.1 | XP_016860336.1 | 9 | 1339 | |||
XM_017004848.1 | XP_016860337.1 | 13 | 1671 | |||
XM_017004849.1 | XP_016860338.1 | 13 | 1671 | |||
XM_017004850.1 | XP_016860339.1 | 13 | 1671 | |||
GenBank mRNA | AK302429.1 | - | 9 | 1220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
