siRNA Details
Gene Symbol: ASB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212556.2 | NP_997721.2 | 2 | 266 | ||
GenBank mRNA | AK123854.1 | - | 1 | 274 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ASB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212556.2 | NP_997721.2 | 2 | 266 | ||
GenBank mRNA | AK123854.1 | - | 1 | 274 |