siRNA Details
Gene Symbol: LOC105377135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534353.2 | XP_011532655.1 | 3 | 2015 |
Gene Symbol: LOC401068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028186.1 | - | 1 | 1145 |
