siRNA Details
Gene Symbol: AMTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286731.1 | NP_001273660.1 | 3, 4 | 238 | ||
NM_212557.3 | NP_997722.1 | 3, 4 | 241 | |||
GenBank mRNA | AB593161.1 | - | 3, 4 | 240 | ||
AY358528.1 | - | 3, 4 | 232 | |||
BC121812.1 | - | 3, 4 | 209 | 40109020 | ||
BC121817.1 | - | 3, 4 | 212 | 40109016 | ||
BF664579.1 | - | 3, 4 | 243 |
