siRNA Details
Gene Symbol: STK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000455.4 | NP_000446.1 | 1 | 936 | ||
XM_005259617.2 | XP_005259674.1 | 1 | 446 | |||
XM_005259618.3 | XP_005259675.1 | 1 | 446 | |||
XM_011528209.1 | XP_011526511.1 | 1 | 446 | |||
GenBank mRNA | AF035625.1 | - | 1 | 159 | ||
AF217978.1 | - | 2 | 782 | |||
BC007981.2 | - | 1 | 271 | 3689780 | ||
BC019334.1 | - | 1 | 92 | 3946005 |
