siRNA Details
Gene Symbol: STK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000455.4 | NP_000446.1 | 1 | 749 | ||
XM_005259617.2 | XP_005259674.1 | 1 | 259 | |||
XM_005259618.3 | XP_005259675.1 | 1 | 259 | |||
XM_011528209.1 | XP_011526511.1 | 1 | 259 | |||
GenBank mRNA | AF217978.1 | - | 2 | 595 | ||
BC007981.2 | - | 1 | 84 | 3689780 |
