siRNA Details
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244644.1 | NP_001231573.1 | 2, 3 | 496 | ||
NM_014043.3 | NP_054762.2 | 3, 4 | 588 | |||
XM_011533576.2 | XP_011531878.1 | 3, 4 | 419 | |||
GenBank mRNA | AF151842.1 | - | 3, 4 | 547 | ||
AK002165.1 | - | 3, 4 | 561 | |||
AK002180.1 | - | 3, 4 | 566 | |||
AK296072.1 | - | 2, 3 | 488 | |||
AK316585.1 | - | 3, 4 | 552 | |||
AL080122.1 | - | 3, 4 | 561 | |||
BC001553.1 | - | 3, 4 | 404 | 3460712 | ||
BI821848.1 | - | 3, 4 | 565 | |||
BM828410.1 | - | 3, 4 | 588 | |||
CR533456.1 | - | 3, 4 | 328 | |||
DA426159.1 | - | 3, 4 | 559 |
