siRNA Details
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 6, 7 | 1457 | ||
NM_001282677.1 | NP_001269606.1 | 6, 7 | 1457 | |||
NM_001282678.1 | NP_001269607.1 | 6, 7 | 1492 | |||
XM_017012319.1 | XP_016867808.1 | 2, 3 | 460 | |||
GenBank mRNA | AB008112.1 | - | 6, 7 | 1428 | ||
AB052090.1 | - | 6, 7 | 1361 | |||
AB052091.1 | - | 6, 7 | 1361 | |||
AB052092.1 | - | 6, 7 | 1361 | |||
AB052093.1 | - | 6, 7 | 1361 | |||
AB052094.1 | - | 6, 7 | 1361 | |||
AF026086.1 | - | 6, 7 | 1421 | |||
AF030356.1 | - | 6, 7 | 1365 | |||
AK292955.1 | - | 6, 7 | 1362 | |||
AK293757.1 | - | 6, 7 | 1400 | |||
AK293848.1 | - | 2, 3 | 262 | |||
AK295686.1 | - | 3, 4 | 401 | |||
BC035575.1 | - | 6, 7 | 1399 | 5498274 |
