siRNA Details
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 14 | 2444 | ||
NM_001282677.1 | NP_001269606.1 | 13 | 2273 | |||
NM_001282678.1 | NP_001269607.1 | 14 | 2479 | |||
XM_017012319.1 | XP_016867808.1 | 9 | 1364 | |||
GenBank mRNA | AB008112.1 | - | 14 | 2415 | ||
AB052090.1 | - | 13 | 2177 | |||
AB052091.1 | - | 14 | 2348 | |||
AB052092.1 | - | 14 | 2348 | |||
AB052093.1 | - | 14 | 2348 | |||
AB052094.1 | - | 14 | 2348 | |||
AB209355.1 | - | 4 | 2024 | |||
AF026086.1 | - | 14 | 2408 | |||
AF030356.1 | - | 14 | 2352 | |||
AK292955.1 | - | 14 | 2349 | |||
AK293757.1 | - | 14 | 2387 | |||
AK295686.1 | - | 11 | 1388 | |||
BC035575.1 | - | 14 | 2386 | 5498274 | ||
DB461678.1 | - | 1 | 118 |
