siRNA Details
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 22, 23 | 3743 | ||
NM_001282677.1 | NP_001269606.1 | 21, 22 | 3572 | |||
NM_001282678.1 | NP_001269607.1 | 22, 23 | 3778 | |||
XM_017012319.1 | XP_016867808.1 | 17, 18 | 2663 | |||
GenBank mRNA | AB008112.1 | - | 22, 23 | 3714 | ||
AB052090.1 | - | 21, 22 | 3476 | |||
AB052091.1 | - | 22, 23 | 3647 | |||
AB052092.1 | - | 22, 23 | 3647 | |||
AB052093.1 | - | 22, 23 | 3647 | |||
AB052094.1 | - | 22, 23 | 3647 | |||
AB209355.1 | - | 10, 11 | 4697 | |||
AF026086.1 | - | 22, 23 | 3707 | |||
AF030356.1 | - | 22, 23 | 3651 | |||
AK292955.1 | - | 22, 23 | 3648 | |||
AK293757.1 | - | 22, 23 | 3686 | |||
AK295686.1 | - | 19, 20 | 2687 | |||
BC035575.1 | - | 22, 23 | 3685 | 5498274 |
