siRNA Details
Gene Symbol: OR51A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004748.1 | NP_001004748.1 | 1 | 405 |
Gene Symbol: OR51A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005329.1 | NP_001005329.1 | 1 | 405 | ||
GenBank mRNA | BC140757.1 | - | 1 | 431 | 9021625 |
