siRNA Details
Gene Symbol: ZNF833P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028594.1 | 3 | 1017 | |||
GenBank mRNA | AK126753.1 | - | 10 | 2400 | ||
BC137336.1 | - | 3 | 867 | 9021334 | ||
BC137337.1 | - | 3 | 867 | 9021335 |
