siRNA Details
Gene Symbol: OR2T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004136.1 | NP_001004136.1 | 1 | 478 | ||
GenBank mRNA | BC136961.1 | - | 1 | 500 | 9020953 | |
BC136962.1 | - | 1 | 500 | 9020954 |
Gene Symbol: OR2T35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001827.1 | NP_001001827.1 | 1 | 475 |
