siRNA Details
Gene Symbol: FAM92A1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003612.2 | 1 | 1390 | |||
GenBank mRNA | BC022019.1 | - | 1 | 1353 | ||
BC047622.1 | - | 1 | 1390 | |||
BC059396.1 | - | 1 | 1389 | |||
BC094692.1 | - | 1 | 1354 |
