siRNA Details
Gene Symbol: C1orf186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007544.3 | NP_001007545.1 | 4 | 767 | ||
XM_005272738.3 | XP_005272795.1 | 4 | 1300 | |||
GenBank mRNA | AK122631.1 | - | 4 | 767 | ||
BC071785.1 | - | 4 | 356 | 4609995 |
