siRNA Details
Gene Symbol: CENPVP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033772.1 | 1 | 728 | |||
GenBank mRNA | AY730278.1 | - | 1 | 728 |
Gene Symbol: CENPVP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033773.1 | 1 | 728 |
