siRNA Details
Gene Symbol: OR2W5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004698.2 | NP_001004698.1 | 1 | 643 | ||
GenBank mRNA | BC137203.1 | - | 1 | 643 | 9021200 | |
BC137204.1 | - | 1 | 643 | 9021201 |
