siRNA Details
Gene Symbol: ZNF806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304449.1 | NP_001291378.1 | 4 | 1766 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304449.1 | NP_001291378.1 | 4 | 1766 |