siRNA Details
Gene Symbol: FRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005217.1 | NP_001005217.1 | 4 | 1194 | ||
NM_001286820.1 | NP_001273749.1 | 4 | 1197 | |||
GenBank mRNA | AY714545.1 | - | 4 | 1194 | ||
BC144572.1 | - | 4 | 1182 | 9053101 |
