siRNA Details
Gene Symbol: DFNB59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042702.3 | NP_001036167.1 | 3 | 704 | ||
XM_005246627.2 | XP_005246684.1 | 3 | 929 | |||
XM_005246628.3 | XP_005246685.1 | 3 | 1075 | |||
XM_005246629.3 | XP_005246686.1 | 3 | 828 | |||
XM_011511247.2 | XP_011509549.1 | 3 | 1075 | |||
XM_011511249.2 | XP_011509551.1 | 2 | 337 | |||
XM_011511250.2 | XP_011509552.1 | 3 | 706 | |||
XM_011511251.1 | XP_011509553.1 | 2 | 154 | |||
XM_017004221.1 | XP_016859710.1 | 3 | 1075 | |||
XM_017004222.1 | XP_016859711.1 | 3 | 1243 | |||
XM_017004223.1 | XP_016859712.1 | 2 | 1278 | |||
XM_017004224.1 | XP_016859713.1 | 2 | 1832 | |||
XM_017004225.1 | XP_016859714.1 | 2 | 414 | |||
GenBank mRNA | BC127902.1 | - | 2 | 380 | 40132882 | |
BC127903.1 | - | 1 | 1091 | 40132890 | ||
BC146938.1 | - | 3 | 480 | 9056866 | ||
DQ365827.1 | - | 3 | 704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
