siRNA Details
Gene Symbol: MS4A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012417.2 | NP_001012417.2 | 5 | 570 | ||
GenBank mRNA | AY324188.1 | - | 5 | 570 | ||
BC140799.1 | - | 5 | 444 | 9021667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
