siRNA Details
Gene Symbol: CXorf40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013845.1 | NP_001013867.1 | 5 | 1080 | ||
XM_005274698.1 | XP_005274755.1 | 5 | 1451 | |||
XM_005274699.1 | XP_005274756.1 | 5 | 1483 | |||
XM_005274700.1 | XP_005274757.1 | 6 | 1629 | |||
XM_005274701.2 | XP_005274758.1 | 5 | 1992 | |||
XM_005274702.3 | XP_005274759.1 | 5 | 1186 | |||
XM_006724826.3 | XP_006724889.1 | 5 | 1205 | |||
XM_011531180.2 | XP_011529482.1 | 5 | 1296 | |||
XM_017029585.1 | XP_016885074.1 | 5 | 1502 | |||
XM_017029586.1 | XP_016885075.1 | 6 | 1648 | |||
XM_017029587.1 | XP_016885076.1 | 6 | 1680 | |||
XM_017029588.1 | XP_016885077.1 | 6 | 2189 | |||
XM_017029589.1 | XP_016885078.1 | 5 | 1286 | |||
GenBank mRNA | BC009523.1 | - | 3 | 686 | 3889701 | |
L43575.1 | - | 5 | 1006 | |||
L43578.1 | - | 4 | 808 | |||
U66042.1 | - | 5 | 1067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
