siRNA Details
Gene Symbol: ZNF704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033723.2 | NP_001028895.1 | 2 | 369 | ||
XM_005251279.4 | XP_005251336.2 | 2 | 1000 | |||
XM_017013725.1 | XP_016869214.1 | 2 | 416 | |||
GenBank mRNA | AK131274.1 | - | 2 | 369 | ||
BC136844.1 | - | 2 | 182 | 9020835 | ||
BC136845.1 | - | 2 | 182 | 9020836 |
