siRNA Details
Gene Symbol: ECSCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077693.3 | NP_001071161.1 | 6, 7 | 452 | ||
NM_001293739.1 | NP_001280668.1 | 6, 7 | 452 | |||
GenBank mRNA | BC146912.1 | - | 6, 7 | 424 | 9056840 | |
BC146913.1 | - | 6, 7 | 424 | 9056841 | ||
BI834795.1 | - | 6, 7 | 402 | |||
DQ462572.1 | - | 6, 7 | 446 | |||
EU025066.1 | - | 6, 7 | 452 |
