siRNA Details
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 5 | 1206 | ||
XM_017012933.1 | XP_016868422.1 | 5 | 1210 |
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 5 | 1210 | ||
XM_017012888.1 | XP_016868377.1 | 5 | 1210 |
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 6 | 1247 | |||
GenBank mRNA | BC045636.1 | - | 6 | 1250 | ||
BC068610.1 | - | 6 | 1250 | |||
BC110795.1 | - | 2 | 514 |
