siRNA Details
Gene Symbol: MIR205HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104548.1 | NP_001098018.1 | 4 | 580 | ||
XM_017002058.1 | XP_016857547.1 | 3 | 675 | |||
GenBank mRNA | AK091113.1 | - | 2 | 3041 | ||
AY320401.1 | - | 1 | 29 | |||
BG674039.1 | - | 5 | 672 |
