siRNA Details
Gene Symbol: MIR205HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104548.1 | NP_001098018.1 | 3 | 524 | ||
XM_017002058.1 | XP_016857547.1 | 2 | 619 | |||
GenBank mRNA | AK091113.1 | - | 1 | 1636 | ||
BG674039.1 | - | 3 | 508 | |||
CV575609.1 | - | 3 | 511 | |||
DA461662.1 | - | 3 | 523 |
